m-Cresol, 99%
m-Cresol, 99%
m-Cresol, 99%
Thermo Scientific Chemicals

m-Cresol, 99%

CAS: 108-39-4 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Glass Bottle
Catalog number 110580010
Price (USD)
171.65
Special offer
Online exclusive
Ends: 28-Dec-2026
202.00
Save 30.35
Each
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
171.65
Special offer
Online exclusive
Ends: 28-Dec-2026
202.00
Save 30.35
Each
Chemical Identifiers
CAS108-39-4
IUPAC Name3-methylphenol
Molecular FormulaC7H8O
InChI KeyRLSSMJSEOOYNOY-UHFFFAOYSA-N
SMILESCC1=CC=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light red
Infrared spectrumConforms
AppearanceMay darken on storage
GC>=98.5 %
Appearance (Form)Viscous liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only