4-Cyanophenol, 99%
4-Cyanophenol, 99%
4-Cyanophenol, 99%
Thermo Scientific Chemicals

4-Cyanophenol, 99%

CAS: 767-00-0 | C7H5NO | 119.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 110830250
Price (USD)
46.65
Special offer
Online exclusive
Ends: 28-Dec-2026
54.50
Save 7.85
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
46.65
Special offer
Online exclusive
Ends: 28-Dec-2026
54.50
Save 7.85
Each
Chemical Identifiers
CAS767-00-0
IUPAC Name4-hydroxybenzonitrile
Molecular FormulaC7H5NO
InChI KeyCVNOWLNNPYYEOH-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige to light grey
Infrared spectrumConforms
Melting point110°C to 113°C
GC>=98.5 %
Sulfated ash=<0.2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only