2-Cyanopyridine, 99%
2-Cyanopyridine, 99%
2-Cyanopyridine, 99%
Thermo Scientific Chemicals

2-Cyanopyridine, 99%

CAS: 100-70-9 | C6H4N2 | 104.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 110851000
Price (USD)
37.90
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
37.90
Each
Chemical Identifiers
CAS100-70-9
IUPAC Namepyridine-2-carbonitrile
Molecular FormulaC6H4N2
InChI KeyFFNVQNRYTPFDDP-UHFFFAOYSA-N
SMILESN#CC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Appearance (Form)Fused solid or liquid
Infrared spectrumConforms
Melting point25°C to 32°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only