1,1-Cyclobutanedicarboxylic acid, 99%
1,1-Cyclobutanedicarboxylic acid, 99%
1,1-Cyclobutanedicarboxylic acid, 99%
Thermo Scientific Chemicals

1,1-Cyclobutanedicarboxylic acid, 99%

CAS: 5445-51-2 | C6H8O4 | 144.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 110920250
Price (USD)
169.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
169.00
Each
Chemical Identifiers
CAS5445-51-2
IUPAC Namecyclobutane-1,1-dicarboxylic acid
Molecular FormulaC6H8O4
InChI KeyCCQPAEQGAVNNIA-UHFFFAOYSA-N
SMILESOC(=O)C1(CCC1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with NaOH98.5 to 101.5 %
Loss on drying=<0.1 % (50°C)
Melting point157°C to 161°C
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only