Cyclobutanone, 98+%
Cyclobutanone, 98+%
Cyclobutanone, 98+%
Thermo Scientific Chemicals

Cyclobutanone, 98+%

CAS: 1191-95-3 | C4H6O | 70.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 110930010
Price (USD)
80.65
Online exclusive
89.80
Save 9.15
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
80.65
Online exclusive
89.80
Save 9.15
Each
Chemical Identifiers
CAS1191-95-3
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4200 to 1.4230 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only