Cycloheptanone, 98+%
Cycloheptanone, 98+%
Cycloheptanone, 98+%
Thermo Scientific Chemicals

Cycloheptanone, 98+%

CAS: 502-42-1 | C7H12O | 112.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 111040250
Price (USD)
42.65
Special offer
Online exclusive
Ends: 28-Dec-2026
49.60
Save 6.95
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 28-Dec-2026
49.60
Save 6.95
Each
Chemical Identifiers
CAS502-42-1
IUPAC Namecycloheptanone
Molecular FormulaC7H12O
InChI KeyCGZZMOTZOONQIA-UHFFFAOYSA-N
SMILESO=C1CCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
GC>=98.0 %
Refractive index1.4595 to 1.4635 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only