Transferrin, Bovine APO Form
Transferrin, Bovine APO Form
Gibco™
Transferrin, Bovine APO Form
Iron-free bovine transferrin for cell culture. It is chromatographically purifed and has an iron content ≤50 μg/g protein. Lyophilized powder.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
111080515 g
1110806625 g
11108032100 g
Catalog number 11108051
Price (USD)
-
Request A Quote
Quantity:
5 g
Iron-free bovine transferrin for cell culture. It is chromatographically purified and has an iron content ≤50 μg/g protein. Lyophilized powder.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell TypeMammalian Cells
Product TypeTransferrin
Quantity5 g
FormSolid
Unit SizeEach