Cyclohexylamine, 99%
Cyclohexylamine, 99%
Cyclohexylamine, 99%
Thermo Scientific Chemicals

Cyclohexylamine, 99%

CAS: 108-91-8 | C6H13N | 99.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 111280010
Price (USD)
53.65
Online exclusive
59.90
Save 6.25
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
53.65
Online exclusive
59.90
Save 6.25
Each
Chemical Identifiers
CAS108-91-8
IUPAC Namecyclohexanamine
Molecular FormulaC6H13N
InChI KeyPAFZNILMFXTMIY-UHFFFAOYSA-N
SMILESNC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.4580 to 1.4600 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only