MEM Non-Essential Amino Acids Solution (100X) - FAQs
MEM Non-Essential Amino Acids Solution (100X) - FAQs
View additional product information for MEM Non-Essential Amino Acids Solution (100X) - FAQs (11140068)
1 product FAQs found
What is the solvent used for MEM Non-Essential Amino Acids Solution (100X)?
The solvent is distilled water.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.