Cyclooctanone, 98%
Cyclooctanone, 98%
Cyclooctanone, 98%
Thermo Scientific Chemicals

Cyclooctanone, 98%

CAS: 502-49-8 | C8H14O | 126.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 111440250
Price (USD)
36.65
Special offer
Online exclusive
Ends: 28-Dec-2026
43.30
Save 6.65
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
36.65
Special offer
Online exclusive
Ends: 28-Dec-2026
43.30
Save 6.65
Each
Chemical Identifiers
CAS502-49-8
IUPAC Namecyclooctanone
Molecular FormulaC8H14O
InChI KeyIIRFCWANHMSDCG-UHFFFAOYSA-N
SMILESO=C1CCCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting crystalline solid
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Colorless to white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only