Cyclopentane, 97%, extra pure
Cyclopentane, 97%, extra pure
Cyclopentane, 97%, extra pure
Thermo Scientific Chemicals

Cyclopentane, 97%, extra pure

Cyclopentane, 97%, C5H10, CAS Number-287-92-3, ciclopentano, cyclopentan, cyclopentanes, cyclopentyl group, hsdb 62, hydrocarbons, cyclic c5 and c6, pentamethylene, t86pb90rnu, unii-t86pb90rnu, zyklopentan, 1L, 96% min. (GC), 05, 19, 50.0 deg.C, CHEBI:23492, White, 0.7500g/mL, 206-016-6, -7 deg.C | CAS: 287-92-3 | C5H10 | 70.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 111480010
Price (USD)
252.65
Online exclusive
280.00
Save 27.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
252.65
Online exclusive
280.00
Save 27.35
Each
Chemical Identifiers
CAS287-92-3
IUPAC Namecyclopentane
Molecular FormulaC5H10
InChI KeyRGSFGYAAUTVSQA-UHFFFAOYSA-N
SMILESC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Residue after evaporation=<0.01 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only