Dynabeads™ CD14
Invitrogen™

Dynabeads™ CD14

Dynabeads CD14 are superparamagnetic beads coupled with an anti-human CD14 antibody that enable easy isolation or depletion of human CD14+ cells directly from any sample, including whole blood, bone marrow, buffy coat, PBMC, and tissue digests.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
11150D2 mL
11149D5 mL
Catalog number 11150D
Price (USD)
736.00
Each
Quantity:
2 mL
Price (USD)
736.00
Each

Dynabeads CD14 are superparamagnetic beads coupled with an anti-human CD14 antibody that enable easy isolation or depletion of human CD14+ cells directly from any sample, including whole blood, bone marrow, buffy coat, peripheral blood mononuclear cells (PBMCs), and tissue digests. Advantages of Dynabeads CD14:

  • Fast and efficient isolation of CD14+ cells from any sample—no columns required
  • High purity, yield, and viability of positively isolated CD14+ cells for molecular downstream assays
  • Fast and efficient depletion of CD14+ cells from any sample

About Dynabeads CD14

Dynabeads CD14 are uniform, superparamagnetic beads (4.5 μm diameter) coated with a primary monoclonal antibody specific for the CD14 membrane antigen mainly expressed mainly on human monocytes. Because of the bead size, Dynabeads CD14 can easily and efficiently isolate or deplete cells from viscous samples such as whole blood and bone marrow in about 30 minutes. Dynabeads CD14 will also afford excellent recovery of high-purity, viable cells for use in downstream molecular studies; for example, those in which cells are to be lysed while still attached to beads, and nucleic acids or proteins further purified. Note that intact cells will not be released from these beads, so if you want to isolate CD14+ cells for cell-based applications, or you need to check your samples with flow cytometry, check out our total human monocyte product offerings.

Magnetic bead to based separation offers easy handling

Dynabeads CD14 is added to the sample under continuous mixing to optimize the binding of the Dynabeads to the target cells. By placing the sample on a magnet, the bead-bound target cells are separated from the rest of the sample in just 1 to 2 minutes. For depletion, remove the supernatant to a new tube for further studies and discard the bead-bound cells. For positive isolation for molecular studies, remove the supernatant and wash the bead-bound cells 2 to 3 times in buffer to get optimal purity. The cells can be lysed while still attached to the beads and the supernatant transferred to a new tube for downstream molecular analysis. Starting samples can be whole blood, buffy coat, bone marrow, PBMC, or tissue digests.

Automation-ready for use with KingFisher purification systems

The product is ideal for high-throughput workflows using KingFisher systems. A user guide and scripts for KingFisher Flex and Apex workflows are available for download on this page.

Learn more about Dynabeads products

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMonocytes
High-throughput CompatibilityYes
No. of CellsProcesses ∼8x 10^8 cells total
Output Viability>95%
Product LineDynabeads
Purity or Quality GradeResearch Grade
Quantity2 mL
Sample TypePBMC, Blood
Shelf Life36 Months (from date of manufacture)
Shipping ConditionRoom Temperature
Starting Material Cell No.1 x 10^7 PBMCs per isolation
Target SpeciesHuman
ColorBrown
Diameter (Metric)4.5 μm
For Use With (Application)Cell Depletion, Positive Isolation for Molecular Applications
MaterialMagnetic Polystyrene
Product TypeAntibody Coated Bead
Unit SizeEach
Contents & Storage
This Product Contains: 2 mL Dynabeads™ CD14 coated with anti-CD14 monoclonal antibody.
Storage Conditions: 2°C to 8°C. Do not freeze.