Cyclopentanone, 99+%, pure
Cyclopentanone, 99+%, pure
Cyclopentanone, 99+%, pure
Thermo Scientific Chemicals

Cyclopentanone, 99+%, pure

CAS: 120-92-3 | C5H8O | 84.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 111532500
Price (USD)
66.65
Online exclusive
74.10
Save 7.45
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
66.65
Online exclusive
74.10
Save 7.45
Each
Chemical Identifiers
CAS120-92-3
IUPAC Namecyclopentanone
Molecular FormulaC5H8O
InChI KeyBGTOWKSIORTVQH-UHFFFAOYSA-N
SMILESO=C1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Refractive index1.4360 to 1.4380 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only