Cyclopentylamine, 99+%
Cyclopentylamine, 99+%
Cyclopentylamine, 99+%
Thermo Scientific Chemicals

Cyclopentylamine, 99+%

CAS: 1003-03-8 | C5H11N | 85.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 111590050
Price (USD)
18.65
Online exclusive
20.40
Save 1.75
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
18.65
Online exclusive
20.40
Save 1.75
Each
Chemical Identifiers
CAS1003-03-8
IUPAC Namecyclopentanamine
Molecular FormulaC5H11N
InChI KeyNISGSNTVMOOSJQ-UHFFFAOYSA-N
SMILESNC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<50 APHA
Infrared spectrumConforms
Water=<0.3 %
Appearance (Form)Clear liquid
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only