Cyclopropyl cyanide, 98%, Thermo Scientific Chemicals
Cyclopropyl cyanide, 98%, Thermo Scientific Chemicals
Cyclopropyl cyanide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopropyl cyanide, 98%, Thermo Scientific Chemicals

CAS: 5500-21-0 | Molecular Formula: C4H5N | Molecular Weight: 67.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
111661000100 gGlass bottle
Catalog number 111661000
Price (USD)
124.65
Special offer
Online exclusive
Ends: 28-Dec-2026
147.00
Save 22.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
124.65
Special offer
Online exclusive
Ends: 28-Dec-2026
147.00
Save 22.35
Each
Chemical Identifiers
CAS5500-21-0
IUPAC Namecyclopropanecarbonitrile
Molecular FormulaC4H5N
InChI KeyAUQDITHEDVOTCU-UHFFFAOYSA-N
SMILESN#CC1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4200 to 1.4220 (20°C, 589 nm)
GC>=97.5 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only