1-Decene, ca. 95%
1-Decene, ca. 95%
1-Decene, ca. 95%
Thermo Scientific Chemicals

1-Decene, ca. 95%

CAS: 872-05-9 | C10H20 | 140.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 111910010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS872-05-9
IUPAC Namedec-1-ene
Molecular FormulaC10H20
InChI KeyAFFLGGQVNFXPEV-UHFFFAOYSA-N
SMILESCCCCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=94 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4200 to 1.4220 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only