Deoxybenzoin, 97%
Deoxybenzoin, 97%
Deoxybenzoin, 97%
Thermo Scientific Chemicals

Deoxybenzoin, 97%

CAS: 451-40-1 | C14H12O | 196.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 111960250
Price (USD)
44.65
Special offer
Online exclusive
Ends: 28-Dec-2026
52.70
Save 8.05
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 28-Dec-2026
52.70
Save 8.05
Each
Chemical Identifiers
CAS451-40-1
IUPAC Name1,2-diphenylethan-1-one
Molecular FormulaC14H12O
InChI KeyOTKCEEWUXHVZQI-UHFFFAOYSA-N
SMILESO=C(CC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystalline powder or crystals
Appearance (Color)White to yellow to light brown
Appearance (Form)and/or chunks
Melting point51°C to 58°C
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only