Search
Expand your CHNS/O analysis capabilities and simplify your workflow with an automated Elemental Analyzer.
| Catalog Number | Type | Includes |
|---|---|---|
| 11206155 | Elemental Analyzer | FlashSmart V CHNS/O (+ MVC) |
| 11206100 | Elemental Analyzer | FlashSmart N/PROTEIN |
| 11206105 | Elemental Analyzer | FlashSmart NC SOIL |
| 11206110 | Elemental Analyzer | FlashSmart CHN |
| 11206115 | Elemental Analyzer | FlashSmart CHNS |
| 11206120 | Elemental Analyzer | FlashSmart CHN/O |
| 11206125 | Elemental Analyzer | FlashSmart CHNS/O |
| 11206130 | Elemental Analyzer | FlashSmart N ORG. |
| 11206135 | Elemental Analyzer | FlashSmart NC ORG. |
| 11206140 | Elemental Analyzer | FlashSmart NCS |
| 11206145 | Elemental Analyzer | FlashSmart N LUBRICANT |
| 11206150 | Elemental Analyzer | FlashSmart V CHN/O (+ MVC) |
| 11206160 | Elemental Analyzer | FlashSmart V CHN/CHN (+ MVC) |
| 11206165 | Elemental Analyzer | FlashSmart V CHNS/CHNS (+ MVC) |
Solve more laboratory challenges, improve workflow, and minimize downtime when performing carbon, hydrogen, nitrogen, sulfur and oxygen (CHNS/O) analyses with the Thermo Scientific™ FlashSmart™ Elemental Analyzers. Based on the modified Dumas Method, the FlashSmart Elemental Analyzer is highly flexible, offering modularity that enables over 20 configurations in one instrument. Besides expanding your analytical capabilities, this analyzer features powerful software that supports automation and precise reports, making the system easier to use from start to finish.
Add automation to your daily organic elemental analysis for a wide range of applications without compromising precision and accuracy.