O-G26/ 1 Swinging Bucket Rotor
Thermo Scientific™
O-G26/ 1 Swinging Bucket Rotor
Use the O-G26/1 Swing-Out Rotor for CL10 rotors and accessories.
Have Questions?
Catalog NumberCapacity
112100144 x 135 mL
Catalog number 11210014
Price (INR)
-
Capacity:
4 x 135 mL
Use the Thermo Scientific™ O-G26/1 Swing-Out Rotor with Thermo Scientific™ CL10 Rotors and Accessories.
Includes: Four buckets
Specifications
DescriptionO-G26/ 1 Swinging Bucket Rotor
For Use With (Equipment)Thermo Scientific CL 10 Centrifuges
Capacity4 x 135 mL
Max. RCF2,647 x g
Max. Speed4,000 rpm
TypeSwinging Bucket Rotor
Unit SizeEach