Fiberlite™ F21-48 x 2 Fixed-Angle Rotor
Actual product may vary
Thermo Scientific™
Fiberlite™ F21-48 x 2 Fixed-Angle Rotor
Support micro-volume protocols using the fixed angle rotor, provided with a biocontainment lid.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
11210351Thermo Scientific Sorvall T1/RT1, Megafuge 11, CL31 and C3i Centrifuges
096-4890115804/5810 Centrifuge
11177523Thermo Scientific Sorvall Legend 23R and MR23i Centrifuges
75003664Thermo Scientific Sorvall Legend XT/XF, Sorvall Legend X1, Sorvall ST 40, Sorvall ST 16, Multifuge X3/X3F, Multifuge X1, Megafuge 40, Megafuge
75006527Thermo Scientific Sorvall Legend T Plus and Multifuge 3 Plus Centrifuges
Catalog number 11210351
Price (USD)
-
For Use With (Equipment):
Thermo Scientific Sorvall T1/RT1, Megafuge 11, CL31 and C3i Centrifuges
Support micro-volume protocols using the Thermo Scientific™ Fiberlite™ F21-48 x 2 Fixed-Angle Rotor.
Fiberlite Rotor:
  • Reduces run-times with higher G-forces achieved with lightweight carbon fiber rotors
  • Ensures structural integrity with corrosion- and fatigue-resistant design
  • Backed by a 15-year warranty
Ordering Information:
Tube Adapters are sold separately in sets of 24; O-ring Refresher Kit (021-489-021) is also available. Contact your sales representative for information.
Compatible with:
Thermo Scientific Sorvall™ Legend™ X1, X3, Sorvall ST 40, Multifuge™ X1, X3, Megafuge 40, SL40 and Eppendorf™ 5810/5804 Series, Sorvall Legend T+, Multifuge 3+ Series, Sorvall Legend 23R, MR23i, Thermo Scientific Nalgene™ bottles and tubes
Note:
†Performance may vary by centrifuge model.
Specifications
Angle45°
DescriptionFiberlite F21-48 x 2 Fixed Angle Rotor for Thermo Scientific Sorvall T1/RT1, Megafuge 11, CL31, C3i Series
For Use With (Equipment)Thermo Scientific Sorvall T1/RT1, Megafuge 11, CL31 and C3i Centrifuges
K Factor455
Max. Tube Diameter11 mm
Max. Tube Length50 mm
Radius (Metric) Min.5.1 cm
Capacity48 x 2 mL
Max. RCF25,055 x g
Max. Speed15,200 rpm
TypeFixed Angle Rotor
Unit SizeEach