1,4-Diaminobutane, 99%
1,4-Diaminobutane, 99%
1,4-Diaminobutane, 99%
Thermo Scientific Chemicals

1,4-Diaminobutane, 99%

CAS: 110-60-1 | C4H12N2 | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 112120025
Price (USD)
751.65
Special offer
Online exclusive
Ends: 28-Dec-2026
884.00
Save 132.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
751.65
Special offer
Online exclusive
Ends: 28-Dec-2026
884.00
Save 132.35
Each
Chemical Identifiers
CAS110-60-1
IUPAC Namebutane-1,4-diamine
Molecular FormulaC4H12N2
InChI KeyKIDHWZJUCRJVML-UHFFFAOYSA-N
SMILESNCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4550 to 1.4590 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only