FLASH 2000 N/Protein Analyzers
Actual product may vary
Thermo Scientific™
FLASH 2000 N/Protein Analyzers
This system provides the ultimate in simplicity, precision, and cost effectiveness for the quantification of protein content in every type of food and beverage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
11230200FLASH 2000 N/Protein +MAS 200R
11230215FLASH 2000 NC Soils + MAS 200R
Catalog number 11230200
Price (USD)
-
Type:
FLASH 2000 N/Protein +MAS 200R
Determine Nitrogen concentration and the relative protein content in every type of food and beverage.
The Thermo Scientific™ FLASH 2000 N/Protein analyzer is an essential tool for both Quality Control and Research/Development laboratories in the food industry, which require precise and accurate determinations, limited cost per analysis, and a 24 hour hard worker able to analyze solid and liquid samples continuously.
Providing a reliable, high-speed, accurate 24/7 analytical solution to a range of industries, this series of flexible, configurable analyzers uses a globally recognized technique endorsed by official organizations worldwide.
An Elemental Analysis Solution for a Range of Industries
With an extraordinary range of applications, the modular design of the FLASH 2000 Series OEAs enables 12 configurations tailored to the fields of application they serve.
  • Organic or inorganic chemistry and pharmaceuticals
  • Petrochemistry and energy
  • Environmental analysis
  • Material characterization
  • Agronomy and marine science
  • Food industry
  • Human and animal samples
  • Isotope analysis
  • Academic
Gain Cost Efficiency and Improve Productivity
The simplicity of the OEA method combined with unparalleled data reproducibility and truly quantitative results are the essence of the sophisticated package of benefits offered by the FLASH 2000 Series OEA instruments.
  • Wide analytical range, enabling extensive applications
  • Versatility and modularity, ensuring that performance meets users' requirements no matter how often those requirements change
  • Accuracy and precision due to the integrated mass flow controller, which enables high stability to deliver precise, accurate results in all sample types and concentrations
  • Ease of use as a result of the unique automated functions, reducing the demands on operators
  • Simplified operation with Thermo Scientific™ Eager Xperience™ software, which minimizes user involvement in set-up
  • Compatibility and automation no matter how high your throughput requirement, with a range of autosamplers that ensures efficient analysis
  • Comprehensive and user-friendly software, a useful and powerful tool tailored for every lab requirement
  • Ground-breaking OEA CookBook (4th edition), which provides a wealth of analysis tips to get optimal results every time
Specifications
IncludesGround-breaking OEA CookBook (4th edition)
For Use With (Application)Determining nitrogen concentration and the relative protein content in every type of food and beverage
TypeFLASH 2000 N/Protein +MAS 200R
Unit Size-