Dibenzothiophene, 98%
Dibenzothiophene, 98%
Dibenzothiophene, 98%
Thermo Scientific Chemicals

Dibenzothiophene, 98%

CAS: 132-65-0 | C12H8S | 184.26 g/mol
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number 112540010
Price (JPY)
-
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS132-65-0
IUPAC Name8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular FormulaC12H8S
InChI KeyIYYZUPMFVPLQIF-UHFFFAOYSA-N
SMILESS1C2=C(C=CC=C2)C2=C1C=CC=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to grey
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Melting point97°C to 101°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only