3,4-Dibenzyloxybenzaldehyde, 99%, Thermo Scientific Chemicals
3,4-Dibenzyloxybenzaldehyde, 99%, Thermo Scientific Chemicals
3,4-Dibenzyloxybenzaldehyde, 99%, Thermo Scientific Chemicals
3,4-Dibenzyloxybenzaldehyde, 99%, Thermo Scientific Chemicals

3,4-Dibenzyloxybenzaldehyde, 99%, Thermo Scientific Chemicals

CAS: 5447-02-9 | C21H18O3 | 318.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 112640050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5447-02-9
IUPAC Name3,4-bis(benzyloxy)benzaldehyde
Molecular FormulaC21H18O3
InChI KeyXDDLXZHBWVFPRG-UHFFFAOYSA-N
SMILESO=CC1=CC=C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point89°C to 93°C
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Appearance (Color)White to yellow to light beige
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only