2,5-Dibromonitrobenzene, 97%
2,5-Dibromonitrobenzene, 97%
2,5-Dibromonitrobenzene, 97%
Thermo Scientific Chemicals

2,5-Dibromonitrobenzene, 97%

CAS: 3460-18-2 | C6H3Br2NO2 | 280.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 112860250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3460-18-2
IUPAC Name1,4-dibromo-2-nitrobenzene
Molecular FormulaC6H3Br2NO2
InChI KeyWRGKKASJBOREMB-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(Br)=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point81°C to 85°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only