alpha,alpha'-Dibromo-o-xylene, 96%
alpha,alpha'-Dibromo-o-xylene, 96%
alpha,alpha'-Dibromo-o-xylene, 96%
Thermo Scientific Chemicals

alpha,alpha'-Dibromo-o-xylene, 96%

CAS: 91-13-4 | C8H8Br2 | 263.95 g/mol
数量:
25 g
100 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 112930250
価格(JPY)
-
数量:
25 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS91-13-4
IUPAC Name1,2-bis(bromomethyl)benzene
Molecular FormulaC8H8Br2
InChI KeyKGKAYWMGPDWLQZ-UHFFFAOYSA-N
SMILESBrCC1=CC=CC=C1CBr
さらに表示
Melting point91°C to 96°C
Appearance (Color)White to brown
Infrared spectrumConforms
GC>=95 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only