What additional supplements are required in Grace's Insect Cell Culture Medium, Unsupplemented to make a "complete" formulation that will support insect cell growth?
Grace's Insect Cell Culture Medium, Unsupplemented requires addition of 3.33 g/L of lactalbumin hydrolysate and 3.33 g/L of yeastolate for the base growth medium. For the complete growth medium that will support insect cell growth, 10% fetal bovine serum must also be added. This complete growth medium is also referred to as TNM-FH.