1,4-Dichloro-2-butyne, 99%
1,4-Dichloro-2-butyne, 99%
1,4-Dichloro-2-butyne, 99%
1,4-Dichloro-2-butyne, 99%
Thermo Scientific Chemicals

1,4-Dichloro-2-butyne, 99%

CAS: 821-10-3 | C4H4Cl2 | 122.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 113281000
Price (EUR)
395,00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
395,00
Each
Chemical Identifiers
CAS821-10-3
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5030 to 1.5060 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only