HLR-6 Swinging Bucket Rotor
HLR-6 Swinging Bucket Rotor
Thermo Scientific™

HLR-6 Swinging Bucket Rotor

Accomodate a variety of blood bags varying in size with Thermo Scientific™ HLR-6 Swinging Bucket Rotor. Adapters ranging from 250mL to 550mL provide capacity flexibility.
Have Questions?
Catalog number 11350TS
also known as 11350
Price (CLP)
-
Perform in-line leukofiltration with the Thermo Scientific™ HLR-6 Swinging Bucket Rotor. Adapters ranging from 250mL to 550mL provide capacity flexibility and the ability to accomodate a variety of blood bags varying in size. Optional microplate carriers accommodate 6 x 3 or standard and 6 x 1 deepwell microplates for greater versatility.
  • Max. Speed/RCF: 5000rpm/7205xg
  • Capacity: 6 x 550mL
  • Incorporates a separate filter holder to preserve blood bag integrity
  • Optimized for soft-spin conditions using FDA and AABB guidelines for the production of leuko-reduced packed red cells and platelet concentrates
Includes:

Rotor cover assembly (11237), rotor buckets w/Pall filter holder, anodized Aluminum (3, 2/pkg., 11351), rotor locking tool (11246), rotor locking stud (11238), balancing discs, rubber (1 pkg., 6/pkg., 00335), speed decal (11009), loctite sealing adhesive (65811), instruction manual (11243)
Specifications
Angle90°
DescriptionHLR-6 Swinging Bucket Rotor
For Use With (Equipment)Thermo Scientific Sorvall RC3BP Plus Low-Speed Centrifuge
K Factor8,355
Min. RCF3,156 x g (Bucket)
Radius (Metric)258 mm Bucket, 185.3 mm Microplate Carrier
Radius (Metric) Min.113 mm
Capacity6 x 550 mL (Bucket), 18 Standard/6 Deepwell (Microplate)
MaterialStainless Steel Rotor Body, Aluminum Bucket
Max. RCF7,205 x g (Bucket), 5,186 x g (Microplate)
Max. Speed5,000 rpm
TypeSwinging Bucket Rotor
Unit SizeEach