2,5-Dichlorophenol, 98%
2,5-Dichlorophenol, 98%
2,5-Dichlorophenol, 98%
Thermo Scientific Chemicals

2,5-Dichlorophenol, 98%

CAS: 583-78-8 | C6H4Cl2O | 163.00 g/mol
Have Questions?
Catalog NumberQuantityPackaging
11358050050 gGlass bottle
Catalog number 113580500
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
43.90
Save 6.25
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
43.90
Save 6.25
Each
Chemical Identifiers
CAS583-78-8
IUPAC Name2,5-dichlorophenol
Molecular FormulaC6H4Cl2O
InChI KeyRANCECPPZPIPNO-UHFFFAOYSA-N
SMILESOC1=CC(Cl)=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals of flakes
Melting point54°C to 58°C
Appearance (Color)White to light brown
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only