Sodium Pyruvate (100 mM) - FAQs
Sodium Pyruvate (100 mM) - FAQs
View additional product information for Sodium Pyruvate (100 mM) - FAQs (11360070)
3 product FAQs found
For how long is Sodium Pyruvate (100 mM) stable at 2-8 degrees C after opening the bottle?
When stored properly at 2-8 degrees C, the product is stable for 12 months from date of shipment
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How is your sodium pyruvate supplied?
Sodium pyruvate (Cat. No. 11360070) is supplied as a liquid supplement for cell culture media at 100 mM (100X) and a final concentration of 11,004 mg/L.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the purpose of sodium pyruvate in media?
Sodium pyruvate serves as an additional energy source for cells in culture. It is often added to low-glucose formulations (1.0 g/L glucose) and is sometimes added to higher-glucose formulations as well. Cells can become “hooked” on sodium pyruvate however, and if it is withdrawn suddenly from the media, they may experience a short lag in growth.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.