4,7-Dichloroquinoline, 98%
4,7-Dichloroquinoline, 98%
4,7-Dichloroquinoline, 98%
Thermo Scientific Chemicals

4,7-Dichloroquinoline, 98%

CAS: 86-98-6 | C9H5Cl2N | 198.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 113750250
Price (USD)
36.10
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
36.10
Each
Chemical Identifiers
CAS86-98-6
IUPAC Name4,7-dichloroquinoline
Molecular FormulaC9H5Cl2N
InChI KeyHXEWMTXDBOQQKO-UHFFFAOYSA-N
SMILESClC1=CC=C2C(Cl)=CC=NC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or crystalline solid
Appearance (Color)White to yellow to brown
Melting point84°C to 88°C
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only