Particle-Certified Glass Containers
Actual product may vary
Particle-Certified Glass Containers
Particle-Certified Glass Containers
Particle-Certified Glass Containers
Particle-Certified Glass Containers
Particle-Certified Glass Containers
Thermo Scientific™

Particle-Certified Glass Containers

Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Product TypeColorNo. per Case
114-250CLP250 mLBoston Round BottleClear24
140-40CLP40 mLContainerClear90
120-02ALP60 mLPacker BottleAmber24
114-060ALP60 mLBoston Round BottleAmber24
114-125ALP125 mLBoston Round BottleAmber12
120-04ALP125 mLPacker BottleAmber12
114-125CTLP125 mLBoston Round BottleClear12
121-08ALP250 mLPacker BottleAmber12
114-250ALP250 mLBoston Round BottleAmber12
131-08CLP
also known as 131-08C/LP
250 mLSample JarClear24
132-16CLP500 mLSample JarClear12
122-16ALP500 mLPacker BottleAmber12
113-500ALP500 mLBoston Round BottleAmber12
112-01ALP1000 mLSample BottleAmber12
133-32CLP1000 mLSample JarClear12
110-80ALP2500 mLContainerAmber6
123-80ALP2500 mLPacker BottleAmber4
111-04ALP4000 mLJugAmber4
Catalog number 114-250CLP
Price (USD)
396.00
Each
Capacity (Metric):
250 mL
Product Type:
Boston Round Bottle
Color:
Clear
No. per Case:
24
Price (USD)
396.00
Each
Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.

Key Features:

  • Class 100/10 glass
  • Certificate of Analysis is included with each shipment
  • Available in clear or amber
  • Assembled with low-shedding polypropylene septa with chemically inert PTFE faced liners that do not contain adhesives
  • Individual bottles double bagged to maintain integrity until point of use
  • 4 L jugs suitable for hazardous shipping in conjunction with combination packaging

Recommended for:

  • Low particle applications
Specifications
Capacity8 oz., 250 mL
Capacity (English)8 oz.
Closure LinerPTFE/Silicone
Closure MaterialPolypropylene, Low-shedding
Closure Size24-414 mm
Closure TypeClosed-top
DescriptionClear Glass Boston Round Bottle with Closed-top Closure, 250 mL
Diameter (English)2.375 in.
Diameter (Metric)60.33 mm
Height (English)5.375 in.
Height (Metric)136.53 mm
No. per Case24
Product TypeBoston Round Bottle
Capacity (Metric)250 mL
ColorClear
MaterialGlass
Unit SizeEach