Diethyl aminomalonate hydrochloride, 98%
Diethyl aminomalonate hydrochloride, 98%
Diethyl aminomalonate hydrochloride, 98%
Thermo Scientific Chemicals

Diethyl aminomalonate hydrochloride, 98%

CAS: 13433-00-6 | C7H13NO4·ClH | 211.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 114041000
Price (USD)
203.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
203.00
Each
Chemical Identifiers
CAS13433-00-6
IUPAC Namehydrogen 1,3-diethyl 2-aminopropanedioate chloride
Molecular FormulaC7H14ClNO4
InChI KeyGLFVNTDRBTZJIY-UHFFFAOYSA-N
SMILES[H+].[Cl-].CCOC(=O)C(N)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=97.5 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only