IPL-41 Insect Medium (1X)
IPL-41 Insect Medium (1X)
IPL-41 Insect Medium (1X)
Gibco™

IPL-41 Insect Medium (1X)

Gibco IPL-41 Insect Medium was originally designed for the growth of Spodeptera frugiperda (Sf9) cells and the expression of recombinantRead more
Have Questions?
Catalog NumberQuantity
114050811000 mL
Catalog number 11405081
Price (USD)
99.25
Each
Quantity:
1000 mL
Customize this product
Price (USD)
99.25
Each
Gibco IPL-41 Insect Medium was originally designed for the growth of Spodeptera frugiperda (Sf9) cells and the expression of recombinant Autographa Californica Nuclear Polyhedrosis Virus (rAcNPV) using the Baculovirus Expression Vector System (BEVS).

This IPL-41 Insect Medium is modified as follows:

WithWithout
• L-glutamine• Tryptose phosphate broth
• Sodium bicarbonate• Yeastolate

IPL-41 Insect Medium contains unique components required for insect cell culture, including malic acid, fumaric acid, and alpha-ketoglutaric acid.

cGMP manufacturing and quality system
Gibco IPL-41 Insect Medium is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Gibco IPL-41 Insect Medium is commonly supplemented with 10% Fetal Bovine Serum (FBS) before use. IPL-41 Insect Medium requires ambient CO2 in order to maintain physiological pH.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineSf21, Sf9
Cell TypeInsect Cell
Inorganic SaltsCalcium
Product LineGibco
Product TypeInsect Cell Medium
Quantity1000 mL
Shipping ConditionRoom Temperature
FormLiquid
With AdditivesGlutamine, Sodium Bicarbonate
Without AdditivesNo Tryptose Phosphate Broth, No Yeastolate
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture