3-Diethylaminopropylamine, 99+%
3-Diethylaminopropylamine, 99+%
3-Diethylaminopropylamine, 99+%
Thermo Scientific Chemicals

3-Diethylaminopropylamine, 99+%

CAS: 104-78-9 | C7H18N2 | 130.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 114090010
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
121.00
Save 18.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
121.00
Save 18.35
Each
Chemical Identifiers
CAS104-78-9
IUPAC Name(3-aminopropyl)diethylamine
Molecular FormulaC7H18N2
InChI KeyQOHMWDJIBGVPIF-UHFFFAOYSA-N
SMILESCCN(CC)CCCN
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Refractive index1.4406 to 1.4426 (20°C, 589 nm)
Appearance (Form)Liquid
Water=<0.3 %
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only