Leibovitz's L-15 Medium, 10 x 500 mL - FAQs
Leibovitz's L-15 Medium, 10 x 500 mL - FAQs
View additional product information for Leibovitz's L-15 Medium - FAQs (11415056)
2 product FAQs found
Is there an alternative for L15 Leibovitz Medium (Cat. Not. 11415056)?
The alternative for L15 Leibovitz Medium (Cat. No. 11415056) is Lebovitz's L-15 Medium, GlutaMAX Supplement (Cat. No. 31415029).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the osmolality of Leibovitz's L-15 Medium?
We do provide osmolality information on the certificate of analysis. All lots of Leibovitz's L-15 Medium will meet the osmolality specification of 300-340 mOsm/kg.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.