Diethylbenzene, Mixture of o-, m- and p-isomers, Sum Min. 95%
Diethylbenzene, Mixture of o-, m- and p-isomers, Sum Min. 95%
Diethylbenzene, Mixture of o-, m- and p-isomers, Sum Min. 95%
Thermo Scientific Chemicals

Diethylbenzene, Mixture of o-, m- and p-isomers, Sum Min. 95%

CAS: 25340-17-4 | C10H14 | 134.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Plastic drum
Catalog number 114160100
Price (USD)
522.00
Each
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
522.00
Each
Chemical Identifiers
CAS25340-17-4
IUPAC Name1,2-diethylbenzene
Molecular FormulaC10H14
InChI KeyKVNYFPKFSJIPBJ-UHFFFAOYSA-N
SMILESCCC1=CC=CC=C1CC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4940 to 1.4960 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
GC>=95.0 % (sum of o-, m-, p-isomers)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only