N,N-Diethylethanolamine, 99%, Thermo Scientific Chemicals
N,N-Diethylethanolamine, 99%, Thermo Scientific Chemicals
N,N-Diethylethanolamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Diethylethanolamine, 99%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
11433010010 LPlastic drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 114330100
Price (USD)
-
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS100-37-8
IUPAC Name2-(diethylamino)ethan-1-ol
Molecular FormulaC6H15NO
InChI KeyBFSVOASYOCHEOV-UHFFFAOYSA-N
SMILESCCN(CC)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<50 APHA
Infrared spectrumConforms
Appearance (Form)Clear liquid
Refractive index1.4400 to 1.4420 (20°C, 589 nm)
Water=<0.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only