Diethyl ethylmalonate, 99%
Diethyl ethylmalonate, 99%
Diethyl ethylmalonate, 99%
Thermo Scientific Chemicals

Diethyl ethylmalonate, 99%

CAS: 133-13-1 | C9H16O4 | 188.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 114365000
Price (USD)
96.90
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
96.90
Each
Chemical Identifiers
CAS133-13-1
IUPAC Name1,3-diethyl 2-ethylpropanedioate
Molecular FormulaC9H16O4
InChI KeyVQAZCUCWHIIFGE-UHFFFAOYSA-N
SMILESCCOC(=O)C(CC)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
Refractive index1.4150 to 1.4170 (20°C, 589 nm)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only