Thermo Scientific Chemicals

N,N-Diethylnicotinamide, 97%, Thermo Scientific Chemicals

Catalog number: 114471000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

N,N-Diethylnicotinamide, 97%, Thermo Scientific Chemicals

Catalog number: 114471000
100 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
59-26-7
IUPAC Name
N,N-diethylpyridine-3-carboxamide
Molecular Formula
C10H14N2O
InChI Key
NCYVXEGFNDZQCU-UHFFFAOYSA-N
SMILES
CCN(CC)C(=O)C1=CC=CN=C1
Appearance (Color)
Clear colorless to yellow
Appearance
May darken on storage
Refractive index
1.5240 to 1.5260 (20°C, 589 nm)
GC
>=96.0 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text