2',4'-Dihydroxyacetophenone, 98%
2',4'-Dihydroxyacetophenone, 98%
2',4'-Dihydroxyacetophenone, 98%
Thermo Scientific Chemicals

2',4'-Dihydroxyacetophenone, 98%

CAS: 89-84-9 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 114805000
Price (USD)
277.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
277.00
Each
Chemical Identifiers
CAS89-84-9
IUPAC Name1-(2,4-dihydroxyphenyl)ethan-1-one
Molecular FormulaC8H8O3
InChI KeySULYEHHGGXARJS-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(O)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point143°C to 147°C
HPLC>=97.5 %
Appearance (Color)Off-white to pink to brown
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only