2',6'-Dihydroxyacetophenone, 97%
2',6'-Dihydroxyacetophenone, 97%
2',6'-Dihydroxyacetophenone, 97%
Thermo Scientific Chemicals

2',6'-Dihydroxyacetophenone, 97%

CAS: 699-83-2 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 114820500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS699-83-2
IUPAC Name1-(2,6-dihydroxyphenyl)ethan-1-one
Molecular FormulaC8H8O3
InChI KeyYPTJKHVBDCRKNF-UHFFFAOYSA-N
SMILESCC(=O)C1=C(O)C=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point155°C to 160°C
Appearance (Form)Powder or chunks
Appearance (Color)Yellow to beige to light brown
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only