3,4-Dihydroxybenzaldehyde, 97%
3,4-Dihydroxybenzaldehyde, 97%
3,4-Dihydroxybenzaldehyde, 97%
Thermo Scientific Chemicals

3,4-Dihydroxybenzaldehyde, 97%

CAS: 139-85-5 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 114850250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS139-85-5
IUPAC Name3,4-dihydroxybenzaldehyde
Molecular FormulaC7H6O3
InChI KeyIBGBGRVKPALMCQ-UHFFFAOYSA-N
SMILESOC1=CC=C(C=O)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=96.0 %
Infrared spectrumConforms
Appearance (Color)Beige to grey-brown
Melting point152°C to 157°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only