3,4-Dihydroxycinnamic acid, 99+%, predominantly trans isomer
3,4-Dihydroxycinnamic acid, 99+%, predominantly trans isomer
3,4-Dihydroxycinnamic acid, 99+%, predominantly trans isomer
3,4-Dihydroxycinnamic acid, 99+%, predominantly trans isomer
Thermo Scientific Chemicals

3,4-Dihydroxycinnamic acid, 99+%, predominantly trans isomer

CAS: 331-39-5 | C9H8O4 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 114931000
Price (EUR)
292,00
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
292,00
Each
Add to cart
Chemical Identifiers
CAS331-39-5
IUPAC Name(2Z)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
Molecular FormulaC9H8O4
InChI KeyQAIPRVGONGVQAS-RQOWECAXSA-N
SMILESOC(=O)\C=C/C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green
Infrared spectrumConforms
Appearance (Form)Powder
Titration with NaOH>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only