1,5-Dihydroxynaphthalene, 97%
1,5-Dihydroxynaphthalene, 97%
1,5-Dihydroxynaphthalene, 97%
Thermo Scientific Chemicals

1,5-Dihydroxynaphthalene, 97%

CAS: 83-56-7 | C10H8O2 | 160.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 115001000
Price (USD)
50.40
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
50.40
Each
Chemical Identifiers
CAS83-56-7
IUPAC Namenaphthalene-1,5-diol
Molecular FormulaC10H8O2
InChI KeyBOKGTLAJQHTOKE-UHFFFAOYSA-N
SMILESOC1=CC=CC2=C(O)C=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey-brown to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only