1,2:3,4-Di-O-isopropylidene-D-galactopyranose, 97%
1,2:3,4-Di-O-isopropylidene-D-galactopyranose, 97%
1,2:3,4-Di-O-isopropylidene-D-galactopyranose, 97%
Thermo Scientific Chemicals

1,2:3,4-Di-O-isopropylidene-D-galactopyranose, 97%

CAS: 4064-06-6 | C12H20O6 | 260.28 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 115240250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4064-06-6
IUPAC Name{4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[7.3.0.0²,⁶]dodecan-8-yl}methanol
Molecular FormulaC12H20O6
InChI KeyPOORJMIIHXHXAV-UHFFFAOYNA-N
SMILESCC1(C)OC2OC(CO)C3OC(C)(C)OC3C2O1
さらに表示
Appearance (Color)Clear colorless to yellow
Appearance (Form)Viscous liquid or paste
Infrared spectrumConforms
Refractive index1.4655 to 1.4685 (20°C, 589 nm)
Specific optical rotation-57° to -61° (20°C, 589 nm) (c=3, CHCl3)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only