Dimethoxymethane, 99.5+%
Dimethoxymethane, 99.5+%
Dimethoxymethane, 99.5+%
Thermo Scientific Chemicals

Dimethoxymethane, 99.5+%

CAS: 109-87-5 | C3H8O2 | 76.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass bottle
Catalog number 115560010
Price (USD)
64.65
Special offer
Online exclusive
Ends: 28-Dec-2026
75.40
Save 10.75
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
64.65
Special offer
Online exclusive
Ends: 28-Dec-2026
75.40
Save 10.75
Each
Chemical Identifiers
CAS109-87-5
IUPAC Namedimethoxymethane
Molecular FormulaC3H8O2
InChI KeyNKDDWNXOKDWJAK-UHFFFAOYSA-N
SMILESCOCOC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.50 %
Infrared spectrumConforms
Water=<0.03 %
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only