(3,4-Dimethoxyphenyl)acetic acid, 99%
(3,4-Dimethoxyphenyl)acetic acid, 99%
(3,4-Dimethoxyphenyl)acetic acid, 99%
Thermo Scientific Chemicals

(3,4-Dimethoxyphenyl)acetic acid, 99%

CAS: 93-40-3 | C10H12O4 | 196.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 115615000
Price (USD)
342.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
342.00
Each
Chemical Identifiers
CAS93-40-3
IUPAC Name2-(3,4-dimethoxyphenyl)acetic acid
Molecular FormulaC10H12O4
InChI KeyWUAXWQRULBZETB-UHFFFAOYSA-N
SMILESCOC1=CC=C(CC(O)=O)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<1 % (K.F.)
Appearance (Color)White to beige
Appearance (Form)Powder
Infrared spectrumConforms
Titration with TBAH>=98.5 % (On dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only