3-Dimethylaminobenzoic acid, 99%
3-Dimethylaminobenzoic acid, 99%
3-Dimethylaminobenzoic acid, 99%
Thermo Scientific Chemicals

3-Dimethylaminobenzoic acid, 99%

CAS: 99-64-9 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 115750250
Price (USD)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS99-64-9
IUPAC Name3-(dimethylamino)benzoic acid
Molecular FormulaC9H11NO2
InChI KeyNEGFNJRAUMCZMY-UHFFFAOYSA-N
SMILESCN(C)C1=CC=CC(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to beige
Infrared spectrumConforms
Melting point150°C to 153°C
Titration with NaOH>=97.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only